Would you like to log in again?
CAPS SNAP SOFT RUB/PTFE BEIGE
Supplier: MicroSolv Technology Corp
Caps^ Autosampler, Snap Top, Soft Polyetheylene, Translucent With 1.0Mm Thick Rubber / Ptfe (Red/Beige) Septa. For Use With 12X32Mm Snap Top Vials. Microsolv Brand. 1000/Case.Microsolv Cat. No.9502C-S80-T-M
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAPS SNAP SOFT RUB/PTFE BEIGE 97052-758 9502C-S80-T-M Case of 1,000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty