Would you like to log in again?
Plastic Vials, Caps and Convenience Kits for LCMS and HPLC
Supplier: MicroSolv
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AQ™ MicroSolv™
9502S-PP-CLEAR 9502S-PP-AM 9532S-0PV-K 9532C-0PV-K 9532C-0XV-K 9532C-0XV 9532S-0P-3CK 9532S-0P-05CP 9532S-0P-05CK 9532S-0XV 9532C-3PC 9532S-MS 9532S-0P-3CP 9572C-MS 9502S-PP-AMB-K 9502S-PP-CLR-K 9572S-MS 9532S-0P-1CP 9532C-MS 9532S-0XV-K 9502S-PP-3CP 9532S-MS-K 9532S-0P-1CK 9572S-MS-EPP 9532S-0PV 9532S-MS-EPP 9572S-PP 9532C-MS-K 9532C-0PV
10804-916PK 550.6 USD InStock
10804-916 10804-672 10790-730 97051-420 97052-824 97052-822 97052-820 97052-688 10804-918 97052-686 97052-684 97052-662 10804-922 10804-844 10804-924 10804-926 10804-826 10804-920 97052-816 97052-814 97052-812 97052-656 10804-670 97052-676 97052-674 97051-416 10805-902 97052-672 97052-650
Plastic Vials, Caps and Convenience Kits for LCMS and HPLC
Vials
Sample safe clear or amber plastic vials for biological, ionic and sensitive analytes.
  • Great alternative to glass in many configurations
  • Reduce or eliminate adsorption
These vials have excellent cost to performance value. Plastic specifications include no extractables with standard HPLC solvents and meets or exceeds demanding lab requirements to have minimal impact on sensitive samples. The bioinert surface is excellent for use with biological compounds such as many proteins, peptides, nucleic acids, ion-chromatography and analysis of PFAS.
Convenient kit packs with plastic vials and caps make it easy to manage and keep your sample vials clean. LCMS vials are made from a special virgin PP that is excellent with aqueous LCMS and chosen when testing biological, ionic or other compounds that interact with glass.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty