Would you like to log in again?
CAPILLARY FUSED SILICA PREMADE
Supplier: MicroSolv Technology Corp
Simplus*Bare Fused Silica Capillaries^ Untreated. 50um ID, 67mm long, 375um OD. Window starts 16.9cm from one end of capillary. Precision cleaved
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAPILLARY FUSED SILICA PREMADE 97054-350 04550-67 Pack of 2 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty