Would you like to log in again?
CAPILLARY CONTROLLED FLOW HIGH
Supplier: MicroSolv Technology Corp
Capillary controlled high flow^ Capillary controlled high flow Coated, high flow, 50um ID by 1meter. Use in Capillary Electrophoresis pack of 1
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAPILLARY CONTROLLED FLOW HIGH 97054-360 04650-HF Pack of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty