Would you like to log in again?
FILTER SF .45D NYLON 25MM 1000
Supplier: MicroSolv Technology Corp
HPLC Syringe type Multi-depth prefilters 25mm^ Polypropylene housing, 0.45um nylon membrane. Color : black with white lettering. Good for viscous or highly particuatled soltuions. Excellent for Environmental and Pharmaceutical labs. Supplied with certificate of analysis
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FILTER SF .45D NYLON 25MM 1000 97054-470 58045-NDX-CASE Case of 1,000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty