Would you like to log in again?
VIALS SILAN SCREW CLEAR 10MM
Supplier: MicroSolv Technology Corp
Basik*10-425mm Screw-Thread Autosampler Vials^ Clear silanized deactivated borosilicate glass, 1.8ml 12x32mm, Screw top wide mouth, Wide mouth
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VIALS SILAN SCREW CLEAR 10MM 97054-564 9503S-0DCV Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty