Would you like to log in again?
PerfectION™ Combined Ion Selective Electrode, METTLER TOLEDO®
Supplier: Mettler-Toledo
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
51344806 51344830 51344703 51344718 51344812 51344730 51344715 51344709 51344800 51344818 51344827 51344712 51344724 51344727 51344815 51344715 51344721 51344812 51344821 51344700 51344803 51344809 51344727 51344818 51344718 51344827 51344806 51344803 51344706 51344703 51344706
97054-628EA 1775 USD InStock
CA11007-326 CA11007-324 97054-628 97054-656 97054-634 97054-632 97054-654 97054-638 CA97054-624 97054-658 CA97054-622 97054-630 97054-652 CA97054-660 97054-650 CA97054-632 97054-666 97054-622 CA97054-652 CA97054-630 97054-664 97054-620 97054-626 97054-648 CA97054-656 97054-668 97054-624 CA97054-650 97054-640 97054-662 97054-660
PerfectION™ Combined Ion Selective Electrode, METTLER TOLEDO®
Electrodes ISE Electrodes
Combined ion-selective electrodes consist of a measuring electrode complete with an integrated reference.
  • Combination electrodes
  • Optimum contact
Click & Clear™ junction provides three advantages such as optimum contact between the electrolyte and measurement solution, even in difficult samples. The electrolyte solution can be made to flow easily through the junction by pressing the electrode head downwards. The junction needs to be cleaned between measurements and the electrolyte can be simply emptied and refilled if necessary.
Ordering information: Each electrode is supplied with one replacement module. The reference electrolytes must be ordered separately.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty