Would you like to log in again?
Headspace Caps, MicroSolv
Supplier: MicroSolv
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
95020-26-S 95025-23-4D 95020-25S-T 95025-03-1S 95025-01-1S 95020-40T-SK 95020-21S-T 95020-40T-S 95025-01-3D 95025-04-1S 95020-45S-CD 95025-14-1S 95025-01-2S 95025-01-4D 95025-26-1D 95025-04-4D 95025-14-2S 95025-14-3D 95025-07-1S 95025-26-4B 95020-50S-W 95020-45S-TK 95025-04-2S 95025-09-1S 95025-08-2S 95025-04-5R 95020-55S-CD 95025-26-4R 95020-50S-W-K 95020-55S-W 95025-02-1S 95025-03-3D 95020-45S-T 95025-08-1S 95025-03-2S 95020-55S-CD-K 95020-55S-W-K 95020-45S-CD-K 95025-00-1S 95020-45S-W 95025-02-2S 95020-45S-W-K 95025-04-3D
97057-242PK 137.85 USD InStock
97057-242 97057-164 97054-928 97057-148 97054-948 97054-926 97057-244 97054-930 97054-974 97054-952 97054-972 97054-934 97054-956 76057-646 97054-912 97054-910 97054-932 76057-644 97054-954 10014-050 97057-176 97054-916 97054-938 97054-936 97054-914 75997-580 97057-232 75997-582 97054-918 97054-942 97054-920 97057-728 97054-940 97054-962 97054-946 97054-924 75997-576 10014-046 97054-922 97054-944 10014-048 75997-578 10014-062
Headspace Caps, MicroSolv
Closures Chromatography Vial Closures
MicroSolv™ headspace caps are available as magnetic and with 4 different septa types to meet needs.
The screw cap eliminates the need for heavy crimping tools and assures that the vials are sealed perfectly every time. The design saves time and money, and ensures a good head space analysis.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty