Would you like to log in again?
CAP HS CRMP SIL/PTFE WH/IV 45D
Supplier: MicroSolv Technology Corp
Head Space aluminum crimp silver Cap^ 20mm with 10mm center hole, silicone white/PTFE ivory septa. 45 durometer, 3.0mm thick (3mil PTFE). 100/pack
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAP HS CRMP SIL/PTFE WH/IV 45D 97054-970 95025-13-1S Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty