Would you like to log in again?
Crimping Tools for Aluminum Caps, MicroSolv™
Supplier: MicroSolv
95025-CRIMP 9502C-40T-CRIMP
97054-986PK 598.76 USD InStock
97054-986 97052-280
Crimping Tools for Aluminum Caps, MicroSolv™
Crimpers, Cappers and Decappers
These manual crimping tools are easy to use.
You can adjust the crimping pressure by using the screw-stop in the handle of the crimping jaws. You can also adjust the crimping height by adjusting the pressure block in the crimper head using an Allen wrench. The crimper handles are Chemical Resistant.
Use with 11mm or 20mm aluminum crimp caps to achieve great seals for autosampler vials. The crimping jaws are made of a special alloy that guarantees long life and has a chemically resistant finish for lab use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty