Would you like to log in again?
CAP SNAP TOP SINGLE SLIT SIL/P
Supplier: MicroSolv Technology Corp
Green Snap Top Cap^ With single preslit 0.04 in thick, white Silicone/PTFE natural septa, For use With 11mm, 12x32mm snap top autosampler vials. Basik brand. 100/pack.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAP SNAP TOP SINGLE SLIT SIL/P 97055-044 9502C-E-35G Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty