Would you like to log in again?
Cogent C8 300™ HPLC Columns, MicroSolv
Supplier: MicroSolv
40008-25P-3M 40008-75P-3M 40008-10P-3M 40008-75P-2-3M 40008-05P-2-3M 40008-10P-2-3M 40008-15P-3M 40008-15P-2-3M
97055-180EA 1350.82 USD InStock
97055-180 97055-172 97055-182 97055-174 97055-184 97055-176 97055-186 97055-178
Cogent C8 300™ HPLC Columns, MicroSolv
Chromatography Columns
Using our proprietary bonding technology, the Cogent C8 300™ has a C8 chain bonded directly to the 5 µm, 300A TYPE-C™ silica surface with two separate points of attachment resulting in silcon-carbon bonds.
  • Extremely long column life
  • Polar and non-polar analytes
  • Increase precision of your results
  • Based on silica hydride
Since there are no siloxane bonds attaching the Ligands, the bonded does not hydrolyze in acidic conditions.
This column is a very stable, efficient and unique HPLC column for macro molecules. Reverse Phase (RP), Normal Phase (NP) and Aqueous Normal Phase (ANP) are all possible with the great wide pore column.
A very powerful tool for bio separations labs working with large proteins, peptides, polysaccharides or tryptic digests looking for longer column life and improved precision in results.
Extremely low pH is possible with this column.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty