Would you like to log in again?
VALVE INJ PEEK/CERAMIC19.5UL
Supplier: MicroSolv Technology Corp
Injection Valve^ PEEK* Analytical 1.0mm flow path 19.5ul volume PEEK* liquid contact ceramic stator. Includes fittings ferrules and wrenches for installation. 1 each
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VALVE INJ PEEK/CERAMIC19.5UL 97055-222 83500-01 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty