Would you like to log in again?
VIAL 4.0ML WO AM 13-SCRW PK100
Supplier: MicroSolv Technology Corp
Autosampler Vials^ Screw top. Basik brand, 4ml, 15x45mm, amber borosilicate glass with write on patch, 13-425 threads. 100/pack
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VIAL 4.0ML WO AM 13-SCRW PK100 97055-384 9504S-E-WAV Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty