Would you like to log in again?
TLC Standard Silica Plate, Phenomenex
Supplier: Phenomenex
T-CSF200200-A
97055-788PK 349.49 USD InStock
97055-788
TLC Standard Silica Plate, Phenomenex
TLC Plates
Standard sized thin-layer chromatography plate, with standard silica treatment and aluminum back
When matching with Claricep flash cartridges, the TLC plates serve as a tool for reaction monitoring and method development based on Rf values for flash chromatography.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty