Would you like to log in again?
FERRULE PTFE FOR 1/16 FF NUT
Supplier: MicroSolv Technology Corp
Ferrule^ For flange free male nuts, used with 1/16th low pressure tubing. Made of PTFE. 10/pack
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FERRULE PTFE FOR 1/16 FF NUT 97055-848 45216-F-PTFE Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty