Would you like to log in again?
Clean-Cut90™ Tubing Cutter for PEEK Tubing, Microsolv Tech Corp
Supplier: MicroSolv
46040-02
97055-850EA 25.72 USD InStock
97055-850
Clean-Cut90™ Tubing Cutter for PEEK Tubing, Microsolv Tech Corp
Cutters
This tool helps to achieve a perpendicular cut of the PEEK tubing due to the fact that the your tubing is held in place while the two faced blade cuts straight down on one side and at an angle on the other side of the tubing.
The locking mechanism of the cutter keeps the blade in a closed position when not in use which helps keep the blade from being dulled.
Ordering information: Replacement blades are available. Supplied with one spare blade and a plastic storage case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty