Would you like to log in again?
TUBING PFA 1/16 X.030 5M PK2
Supplier: MicroSolv Technology Corp
Tubing^ PFA (PerFlourAlkoxy) 1/16thin x .030in (0.75mm) ID. Low pressure 5 meter roll
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBING PFA 1/16 X.030 5M PK2 97055-886 48310-30 Pack of 2 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty