Would you like to log in again?
EXP®2 Stem Trap Kits and Replacement Stems, 3.0 µm EXP
Supplier: Optimize Technologies
EXP® EXP®2
15-03997-HK 15-04009-HL 15-04009-HJ 15-04001-HI 15-04014-HK 15-04014-HI 15-04014-HG 15-04009-HK 15-04009-HI 15-04021-HL 15-03997-HL 15-04021-HH 15-03992-HJ 15-03992-HL 15-03992-HH 15-04014-HH 15-04009-HM 15-04020-HJ 15-04021-HG 15-04014-HL 15-04020-HI 15-04003-HJ 15-04008-HJ 15-04003-HH 15-03992-HG 15-04009-HG 15-04020-HG 15-04003-HK 15-04003-HM 15-04020-HM 15-04008-HI 15-04015-HI 15-04015-HJ 15-04001-HG 15-04003-HL 15-04015-HK 15-04003-HG 15-04021-HK 15-03992-HM 15-04001-HL 15-04015-HG 15-04021-HM 15-04008-HG 15-04009-HH 15-03997-HH 15-03992-HI 15-04020-HH 15-04020-HL 15-04001-HH 15-04001-HM 15-04008-HL 15-04015-HL 15-04009-HL 15-04021-HJ 15-03992-HK 15-03997-HG 15-04008-HM 15-03997-HM 15-04014-HJ 15-04015-HH 15-04021-HI 15-04001-HJ 15-04015-HM 15-03997-HJ 15-04020-HK 15-04001-HK 15-04003-HI 15-04014-HM 15-03997-HI 15-04008-HK 15-04008-HH
97056-080PK 1256.35 USD InStock
97056-080 97056-122 97056-364 97056-088 97056-362 97056-120 97056-086 97056-360 97056-084 97056-082 97056-128 97056-402 97056-126 97056-368 97056-400 97056-124 97056-366 97056-408 97056-406 97056-092 97056-090 97056-132 97056-374 97056-098 97056-130 97056-372 97056-096 97056-370 97056-094 97056-416 97056-414 97056-138 97056-412 97056-378 97056-136 97056-410 97056-376 97056-134 97056-418 97056-386 97056-100 97056-144 97056-384 97056-142 97056-382 97056-140 97056-380 97056-108 97056-348 97056-106 97056-148 97056-104 97056-346 97056-388 97056-146 97056-102 97056-396 97056-352 97056-110 97056-394 97056-350 97056-392 97056-390 97056-118 97056-358 97056-116 97056-356 97056-114 97056-354 97056-112 97056-398
EXP®2 Stem Trap Kits and Replacement Stems, 3.0 µm EXP
Chromatography Columns
The entire EXP®2 stem trap and reusable holder are only slightly larger than a standard HPLC fitting. Its slim architecture allows it to easily fit into crowded instrument compartments or to connect directly to tightly-spaced injection ports.
  • Rated to 20000+ psi (1400+ bar)
  • Hand-tight and wrench-tight configurationwith auto-adjusting ZDV connection
  • Low-volume/low-dispersion cartridges
  • For nano flow applications
When tightened by hand, the EXP2 stem trap seals to 8700+ psi. All configurations incorporate wrench-flats to enable flawless sealing to 20000+ psi (1400+ bar). The unique packed floating stem installs directly into any 10-32 port and automatically adjusts to provide a perfect ZDV connection. The EXP system enables chemists to quickly remove detergents or salts which can affect the ionization process in MS work. This trapping technique can concentrate the sample directly on-line, allowing for increased recovery of precious sample material compared to off-line techniques. On-line trapping readily lends itself to automation for high-throughput analysis in UHPLC/MS applications. Specialized features, patented technology, precision engineering and state-of-the-art manufacturing make the new EXP2 stem trap an unbeatable choice for ultra high-pressure nano-flow trapping applications.
The new EXP2 design incorporates integral ferrule into the front-end of the holder. No separate ferrule to lose.
Ordering information: Custom packing available.
Delivery information: Kit includes EXP®2 stem holder with fittings and 3 EXP® stem traps (with bed volumes from 0.17 to 2.6 µl), intended for many repeat uses.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty