The entire EXP®2 stem trap and reusable holder are only slightly larger than a standard HPLC fitting. Its slim architecture allows it to easily fit into crowded instrument compartments or to connect directly to tightly-spaced injection ports.
- Rated to 20000+ psi (1400+ bar)
- Hand-tight and wrench-tight configurationwith auto-adjusting ZDV connection
- Low-volume/low-dispersion cartridges
- For nano flow applications
When tightened by hand, the EXP2 stem trap seals to 8700+ psi. All configurations incorporate wrench-flats to enable flawless sealing to 20000+ psi (1400+ bar). The unique packed floating stem installs directly into any 10-32 port and automatically adjusts to provide a perfect ZDV connection. The EXP system enables chemists to quickly remove detergents or salts which can affect the ionization process in MS work. This trapping technique can concentrate the sample directly on-line, allowing for increased recovery of precious sample material compared to off-line techniques. On-line trapping readily lends itself to automation for high-throughput analysis in UHPLC/MS applications. Specialized features, patented technology, precision engineering and state-of-the-art manufacturing make the new EXP2 stem trap an unbeatable choice for ultra high-pressure nano-flow trapping applications. The new EXP2 design incorporates integral ferrule into the front-end of the holder. No separate ferrule to lose.
Ordering information: Custom packing available.
Delivery information: Kit includes EXP®2 stem holder with fittings and 3 EXP® stem traps (with bed volumes from 0.17 to 2.6 µl), intended for many repeat uses.