Would you like to log in again?
Accessories for EXP®2 Stem Trap Kits and Replacement Stems, 2.7 µm HALO®
Supplier: Optimize Technologies
EXP®2
15-02-03996
97056-472EA 427.27 USD InStock
97056-472
Accessories for EXP®2 Stem Trap Kits and Replacement Stems, 2.7 µm HALO®
Chromatography Columns
The reusable holder are only slightly larger than a standard HPLC fitting. The slim architecture allows it to easily fit into crowded instrument compartments or
to connect directly to tightly-spaced injection ports.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty