Would you like to log in again?
EXP®2 Stem Trap Kits and Replacement Stems, 2.7 µm HALO®
Supplier: Optimize Technologies
EXP®2
15-03992-HA 15-03992-HF 15-04015-HA 15-04001-HA 15-03997-HF 15-03992-HN 15-04001-HB 15-04015-HN 15-04009-HF 15-03997-HD 15-04008-HB 15-04015-HB 15-03997-HA 15-04003-HF 15-04014-HB 15-04008-HA 15-03997-HN 15-04008-HE 15-04003-HD 15-04015-HE 15-04020-HA 15-04014-HN 15-04020-HE 15-03992-HB 15-04001-HF 15-04015-HD 15-04009-HB 15-04009-HA 15-04021-HB 15-04021-HF 15-04021-HD 15-04020-HF 15-04001-HE 15-04001-HD 15-04009-HN 15-04020-HD 15-04009-HE 15-04020-HN 15-04008-HD 15-04021-HN 15-04014-HE 15-03992-HE 15-04014-HF 15-04003-HB 15-04008-HN 15-04020-HB 15-04014-HA 15-04021-HE 15-04009-HD 15-04003-HA 15-04008-HF 15-04003-HN 15-03997-HB 15-04021-HA 15-03992-HD 15-04001-HN 15-03997-HE 15-04014-HD 15-04015-HF 15-04003-HE
97056-484PK 1517.74 USD InStock
97056-484 97056-440 97056-482 97056-480 89208-100 97056-448 89208-102 97056-446 97056-488 97056-444 97056-486 97056-442 89208-092 97056-452 97056-496 97056-450 89208-094 97056-494 97056-492 89208-090 97056-490 97056-458 97056-456 89208-096 89208-098 97056-454 97056-498 97056-462 97056-460 97056-504 89208-088 97056-426 97056-502 97056-424 97056-468 97056-500 89208-084 97056-466 97056-422 89208-086 97056-420 97056-464 89208-108 97056-508 89208-104 89208-106 97056-506 97056-428 97056-474 97056-430 97056-438 89208-110 97056-436 97056-512 89208-112 97056-434 97056-478 97056-510 97056-476 97056-432
EXP®2 Stem Trap Kits and Replacement Stems, 2.7 µm HALO®
Chromatography Columns
The entire EXP®2 stem trap and reusable holder are only slightly larger than a standard HPLC fitting. Its slim architecture allows it to easily fit into crowded instrument compartments or to connect directly to tightly-spaced injection ports.
  • Rated to 20000+ psi (1400+ bar)
  • Hand-tight and wrench-tight configurationwith auto-adjusting ZDV connection
  • Low-volume/low-dispersion cartridges
  • For nano flow applications
When tightened by hand, the EXP2 stem trap seals to 8700+ psi. All configurations incorporate wrench-flats to enable flawless sealing to 20000+ psi (1400+ bar). The unique packed floating stem installs directly into any 10-32 port and automatically adjusts to provide a perfect ZDV connection. The EXP system enables chemists to quickly remove detergents or salts which can affect the ionization process in MS work. This trapping technique can concentrate the sample directly on-line, allowing for increased recovery of precious sample material compared to off-line techniques. On-line trapping readily lends itself to automation for high-throughput analysis in UHPLC/MS applications. Specialized features, patented technology, precision engineering and state-of-the-art manufacturing make the new EXP2 stem trap an unbeatable choice for ultra high-pressure nano-flow trapping applications. The new EXP2 design incorporates integral ferrule into the front-end of the holder. No separate ferrule to lose.
Ordering information: Custom packing available.
Delivery information: Kit includes EXP®2 stem holder with fittings and 3 EXP® stem traps (with bed volumes from 0.17 to 2.6 µl), intended for many repeat uses.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty