Would you like to log in again?
TEE PEEK .010 BODY ONLY
Supplier: MicroSolv Technology Corp
Tee Adapter^ HPLC, PEEK, 0.010 (0.3mm) bore with 10-32 threads. Male nuts sold separately
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TEE PEEK .010 BODY ONLY 97056-784 49302-T10 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty