Would you like to log in again?
TUBING PEEK 010 ID 50MM BL
Supplier: MicroSolv Technology Corp
Tubing, HPLC, Peek^ coiled, 0.010in ID x 16thin OD 5000 psi max pressure 50mm resting 300mm extended coil total Length: 1585mm w/male nuts. Blue
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBING PEEK 010 ID 50MM BL 97056-880 49308-501B Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty