Would you like to log in again?
TUBING PRECUT .005X20CM RED
Supplier: MicroSolv Technology Corp
Tubing, Pre-cut Stainless Steel^ For HPLC, 0.005 ID x 1/16in x 20cm long. Color coded red.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBING PRECUT .005X20CM RED 97056-904 49405-20 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty