Would you like to log in again?
TUBING PEEK 1/16X007 SUPERT
Supplier: MicroSolv Technology Corp
Tubing, Super-T, HPLC Peek^ 0.007 ID (0.064mm) x 1/16th OD, ultra high tolerance. Color coded yellow double stripe. 5 meter roll.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBING PEEK 1/16X007 SUPERT 97057-022 47305-ST-07 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty