Would you like to log in again?
CAPS SNAPTP TRANS PRECUT (Y)
Supplier: MicroSolv Technology Corp
Caps, Snap top^ With partial precut (Y) silicone/PTFE septa fitted in the cap. Transparent. For use with 12x32mm snap crimp vials
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAPS SNAPTP TRANS PRECUT (Y) 97057-170 9502C-34CT Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty