Would you like to log in again?
VIALS 10ML 46 X 23MM CRP CL BL
Supplier: MicroSolv Technology Corp
Vials, Head Space^ 20mm, crimp top, 10ml, clear, long neck flat bottom, 46mm x 223mm. Use with Agilent, Dani Fisons autosamplers. Basik brand.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VIALS 10ML 46 X 23MM CRP CL BL 97057-184 95025-EAG10C Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty