Would you like to log in again?
EPP 10ML CENTER DRAINING VIAL
Supplier: MicroSolv Technology Corp
Easy Purchase Pack Vials & Caps^ 10ml center draining, 20.75 x 61mm, 18-400 clear screw top vials & polyproylene caps with bonded silicone / PTFE septa.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
EPP 10ML CENTER DRAINING VIAL 97057-822 98110-1CP Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty