Would you like to log in again?
EPP STORAGE VIALS/SCREW CAPS
Supplier: MicroSolv Technology Corp
Easy Purchase Pack Vials^ Storage, 8ml clear borosilicate glass (61x16.6mm) w/ black screw caps w/ butyl rubber/PTFE fitted liner. Solid top.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
EPP STORAGE VIALS/SCREW CAPS 97057-842 97100-08-EPP Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty