Would you like to log in again?
LASTDROP FILTER 10.0UM PTF1/16
Supplier: MicroSolv Technology Corp
Filter^ HPLC mobile phase inlet, Last Drop type, 10.0um PTFE filter with 1/16th tubing connector.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LASTDROP FILTER 10.0UM PTF1/16 97057-886 58846-08 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty