Would you like to log in again?
MALE NUTS SS LONG 10PK
Supplier: MicroSolv Technology Corp
Male Nut^ HPLC, 316L stainless steel. 10-32 hex head for use on extra long Rheodyne threads.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MALE NUTS SS LONG 10PK 97057-926 49590-03 Pack of 2 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty