Would you like to log in again?
MALE NUT 2PC DUBL FERRULE PEEK
Supplier: MicroSolv Technology Corp
Male Nuts^ HPLC, Peek, 2 piece, for use with double ended ferrule, 12mm knurled head, total length 18mm, 10mm thread length, 10-32, natural color. Ferrules sold separately.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MALE NUT 2PC DUBL FERRULE PEEK 97057-992 47320-31 Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty