Would you like to log in again?
NUTS LP PP BLACK 1/16 MALE
Supplier: MicroSolv Technology Corp
Male Nuts^ Low pressure, polypropylene, black, use w/ 1/16thin flanged tubing. 1/4-28 hreads. Washers sold separately.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
NUTS LP PP BLACK 1/16 MALE 97058-024 45016-K Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty