Would you like to log in again?
LOOP SAMPLE PEEK VALCO 2UL
Supplier: MicroSolv Technology Corp
Sample Loop^ HPLC, 2ul volume, Super-T PEEK tubing 0.06mm ID, use with Valco injectors.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LOOP SAMPLE PEEK VALCO 2UL 97058-082 47002-002 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty