Would you like to log in again?
WASHER FOR FLANGED NUTS 1/16TH
Supplier: MicroSolv Technology Corp
Washers For Male Nuts^ Low pressure, polypropylene, for use w/ 1/16thin flanged tubing.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
WASHER FOR FLANGED NUTS 1/16TH 97058-142 45016-WASHER Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty