Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Digital Vortex Genie® 2 For Applied Biosystems® Mixer Shaker, Scientific Industries

Supplier: Scientific Industries

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

SI-A536
97060-142EA 644.09 USD InStock
97060-142
Digital Vortex Genie® 2 For Applied Biosystems® Mixer Shaker, Scientific Industries
Shakers and Mixers Vortex Mixers
Specifically configured Digital Vortex-Genie® 2 with microplate accessory for Applied Biosystems® Microplate Protocols Only, includes a specially designed adaptor

Certifications: CE marked.

  • Specifically configured
  • Digital control
  • Variable speed

Digital control, display of speed and time provide accuracy, reproducibility, and repeatability. Variable speed allows for slow shaking up to high vortexing. Timing function incorporated for Hands-Free mixing 1 to 99 minutes or continuous. May be used in cold rooms or incubators. (Does not include 3" Platform and Pop-Off Cup).

Certifications: CE marked.

Ordering information: Microplate adaptor included. The 230 V model is supplied without a plug.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty