Would you like to log in again?
Orbital-Genie™ Large Capacity Orbital Shakers, 230 V
Supplier: Scientific Industries
Genie
SI-1601 SI-1501 SI-1701
97060-148EA 6089.06 USD InStock
97060-148 97060-156 97060-152
Orbital-Genie™ Large Capacity Orbital Shakers, 230 V
Shakers and Mixers Orbital Shakers
The Orbital-Genie™ Large Capacity Orbital Shaker is a powerful large capacity (460×460 mm platform) orbital shaker designed to hold any type of container.
  • 230 V model
  • Holds any type of container
  • Fully programmable
  • Variable speed
  • Three platform configurations
Fully programmable shaker features variable speed including orbit reverse, timer from 1 minute to 99 hours with alarm, programmable acceleration and deceleration rates, programmable ON/OFF times for interval shaking. Operator can create any mixing or shaking action from smooth vortex to sloshing aeration. Available in three platform configurations: Universal Ratcheting Clamps - provide the flexibility for using a mix of different size and shape vessels on a single platform. Adhering Mat - provides a flat platform with adhering rubber mat perfect for use with open containers, especially large containers. Standard Fask Clamps - is equipped with a platform for accepting various size flasks or beakers. (Clamps sold separately). All units can be used in a cold room or incubator and CO₂ environments.
Certifications: CE marked.
Ordering information: Two ratcheting and one non-ratcheting cushioned clamps included with Ratcheting Clamps model. The 230 V model is supplied without a plug.
Caution: 230 V models. Corresponding models with voltage suitable for use in US (97060-146, 97060-150, 97060-154) are also available, please see separate product page.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty