Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vortex Genie® Mixers, Scientific Industries

Supplier: Scientific Industries

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Genie®
SI-0511 SI-0513 504-0039-00 SI-V203 146-3011-00 SI-H512 504-0232-00 504-0234-00 0K-0500-902 SIV209 504-0233-00 SI-0570 146-6005-00 SI-0136 503-0283-00 SI-V206 SI-H524 503-0278-00 SI-V525 0K-0500-901 503-0280-00 SI-H506 0K-0513-900 504-0235-00 SI-V512 SI-V203 SI-T236 SI-0246 0K-0236-408 SI-0236 SI-0510 OK-0500-901 SI-0564 SI-V209 568-0001-00 SI-A236 SIV206 503-0279-00 503-0277-00 SI-V524 SI-V506 503-0282-00 580-2013-00
14216-188EA 606.34 USD InStock
14216-188 58815-218 490014-806 58815-216 490010-036 97060-186 89415-150 97060-184 490010-018 14216-184 97060-182 490010-038 490010-016 13502-390 89202-338 97060-180 58815-180 58815-222 58815-188 58815-220 58815-186 14221-972 58815-185 58815-184 490010-040 58815-206 13502-386 13502-388 58815-224 97060-176 89202-322 490010-026 97060-174 89401-418 89401-416 89037-464 89401-420 97060-178 58815-212 58815-234 58815-210 13500-962 58815-230
Vortex Genie® Mixers, Scientific Industries
Shakers and Mixers Vortex Mixers
Vortex Genie® Mixers are available in multiple models and with a vast array of available platforms and accessories.

  • Available with a vast array of accessories
  • Digital Vortex-Genie features digital time and speed setting, touch or continuous mode
  • Two-year manufacturer's warranty

The Vortex Genie 1 touch mixer provides stronger handheld vortexing.

Optional accessories vastly increase the versatility of the vortexers. The TurboMix™ attachment provides hands-free, simultaneous, extremely vigorous vortexing and mixing of up to 12 microtubes. It is designed for cell disruptions and other glass bead procedures and pellet resuspensions. The Multiple Sample Starter Set accessory includes a 15.2 cm (6") platform with one 60-microtube insert for 0.5–2.0 ml tubes and one microplate insert. The Large Sample Set accessory includes a recessed platform and two elastic bands for securing large flasks and beakers up to 500 ml. Also includes one foam insert each for 9–16 mm test tubes, 16–29 mm test tubes, 29–37 mm test tubes, and one customizable blank foam insert. The Two-Tier Insert for Microplate accessory fits into the 15.2 cm (6") Platform Head Only.

Economical and easy-to-use, it features violent high-speed vortexing action for maximum agitation in problem applications. Operator simply holds the container on the mixing head and the mixer starts automatically at maximum speed. Stops on release. The familiar Vortex Genie 2 mixers accept a large range of snap-on accessories for variable speed mixing, from gentle shaking to violent vortexing of most containers up to 500 ml. Select touch-on or continuous operation. The timed Vortex Genie 2T mixer includes a timer function in either continuous untimed, touch, or hands-free mode. Touch mode: 1–60 seconds. Hands-free mode: 1–60 minutes. The Digital Vortex Genie 2 mixer is ideal for applications that require vortexing/shaking of various types/sizes of tubes, vials, bottles, or plates, includes digital control and display of speed, time, accuracy, reproducibility, and repeatability. Touch mode: 1–99 seconds. Hands-free mode: 1–99 minutes. All models are built of a solid 4 kg (8.8 lbs.) cast-metal base with rubber feet that prevent “walking.”

Certifications: CE marked. cETLus, ETL listed.

Ordering information: Additional Vortex-Genie accessories (sold separately) provide for hands-free operation and increased throughput.

Delivery information: Supplied with manufacturer’s two-year warranty on all models. The Vortex Genie 1 is supplied with a quick change standard pop-off cup to accommodate single test tubes. The Vortex Genie 2, 2T, and digital models are supplied with a quick-change standard cup for single test tubes and a 7.6 cm (3") platform for beakers, flasks, and multiple test tubes.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty