Would you like to log in again?
MRS Agar Plate, Teknova
Supplier: Teknova
M1350 M1350 M1350
97060-361EA 95.46 USD InStock
97060-361 97060-360 CAM1350
MRS Agar Plate, Teknova
Media for Microbiology Prepared Media for Microbiology
The plate contains agar media.
The plate contains agar media made of 1% tryptone, 1% beef extract, 0.5% yeast Eextract, 2% glucose, 0.5% sodium acetate, 0.2% dipotassium phosphate, 0.2% ammonium citrate, 0.01% magnesium sulfate, 0.005% manganese sulfate, 0.1% tween-80 and 2.1% agar at pH 6.5. Plates are to be stored at room temperature.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty