Would you like to log in again?
VWR® HistoChoice® Clearing Agent
Supplier: Avantor
HistoChoice®
H103-1L H103-4L
97060-932EA 45.51 USD InStock
97060-932 97060-934
VWR® HistoChoice® Clearing Agent
Stains Microscopy and staining reagents
Less hazardous replacement for xylene and toluene. Compatible with organic mounting media, clearing agent is designed to work with isopropanol in automated processors. It also offers faster wax dissolution and slower evaporation than xylene with no oily residue. Clearing agent can be recycled by distillation.
  • Less toxic replacement for xylene
  • Virtually odorless
  • Faster wax dissolution and slower evaporation than xylene
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty