Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EDTA disodium salt dihydrate for biotechnology

Supplier: VWR
Warning
Synonyms: Ethylenediaminetetra-acetic acid disodium salt dihydrateEdetate disodium dihydrateEthylenediaminetetraacetic acid, disodium salt, dihydrateKestranal® 2S 2 H2OEthylenedinitrilotetra-acetic acid disodium salt dihydrateTitriplex IIIKestranal® 2S dihydrateEdetic acid disodium salt dihydrateDisodium dihydrogen ethylenediaminetetraacetate dihydrate

0105-12KG 0105-1KG 0105-2.5KG 0105-500G
97061-016EA 1411.69 USD InStock
97061-016 97061-018 97061-022 97061-020
EDTA disodium salt dihydrate for biotechnology
EDTA disodium salt dihydrate
A reversible metalloprotease inhibitor useful for chelating metal ions.


Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Melting Pt: 255 °C
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://us-legacy.vwr.com/stibo/web/std.lang.all/99/56/2299956.jpg
Order Now
Specification Test Results
DNase none detected
Heavy Metals (as Pb) = 0.005%
Insolubles = 0.005%
Iron = 0.01%
Loss on Drying 8.7% - 11.4%
Nitrilotriacetic Acid = 0.1%
pH (5%, Water) @25°C 4.0 - 6.0
Protease none detected
Purity = 99%
RNase none detected

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty