Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

AMP (2-Amino-2-methylpropanol), high purity

Supplier: VWR
Warning
Synonyms: 2-Amino-2-methyl-1-propanolAMP

0136-4L
97061-064EA 243.13 USD InStock
97061-064
AMP (2-Amino-2-methylpropanol), high purity
AMP (2-Amino-2-methylpropanol)
5% water has been added to maintain liquidity at room temperature.


Formula: (CH₃)₂C(NH₂)CH₂OH
MW: 89.14 g/mol
Boiling Pt: 165 °C (1013 hPa)
Melting Pt: 18…26 °C
Density: 0.667 g/cm³ (20 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00008051
CAS Number: 124-68-5
EINECS: 204-709-8
Merck Index: 13,00448
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty