Synonyms:
Diammonium sulfate
, Sulphuric acid diammonium salt
, Diammonium sulphate
, Sulfuric acid diammonium salt
0191-1KG
0191-5KG
0191-50KG
0191-12KG
0191-10KG
97061-176EA
394.83
USD
InStock
97061-176
97061-178
97061-184
97061-180
97061-182
Ammonium sulfate, Ultrapure
Ammonium sulphate
May be used for the precipitation or fractionation of proteins or for purification of antibodies. Useful for crystallographic analysis of nucleic acids and proteins.