Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Aspartic acid ≥98%, powder, high purity

Supplier: VWR
Synonyms: L-α-Aminosuccinic acid(S)-2-Aminosuccinic acidD(S)-(+)-Aspartic acidL(+)-2-Amino-1,4-butanedioic acidH-Asp-OHL-αlpha-Aminosuccinic acid(S)-(+)-2-Amino-1,4-butanedioic acidAsp

0192-500G
97061-188CS 708.13 USD InStock
97061-188
L(+)-Aspartic acid ≥98%, powder, high purity
L(+)-Aspartic acid
Non-animal origin. White powder


Formula: C₄H₇NO₄
MW: 133.1 g/mol
Boiling Pt: 311 °C (1013 hPa)
Melting Pt: 251 °C
Density: 1.6613 g/cm³ (13 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002616
CAS Number: 56-84-8
EINECS: 200-291-6
Merck Index: 13,00845
Order Now
Specification Test Results
Loss on Drying = 0.2 %
Purity = 98 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty