Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Fructose, powder, high purity

Supplier: VWR
Synonyms: D(-)-Levuloseβ-D-(-)-Fructose(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-oneFructose

0226-12KG 0226-1KG 0226-5KG
97061-236CS 186.59 USD InStock
97061-236 97061-242 97061-234
D-(-)-Fructose, powder, high purity
D(-)-Fructose
Fine white powder.


Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 552 °C (1013 hPa)
Melting Pt: 100…110 °C
Density: 1.59 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00148910
CAS Number: 57-48-7
EINECS: 200-333-3
Merck Index: 13,04295
Order Now
Specification Test Results
Arsenic = 0.0001%
Calcium & Magnesium = 0.005%
Chloride = 0.018%
Heavy Metals (as Pb) = 0.0005%
Loss on Drying = 0.5%
Purity = 99%
Residue on Ignition = 0.5%
Sulfate = 0.025%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty