Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Manganese(II) sulfate monohydrate, Reagent Grade

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Warning
Synonyms: Manganous sulfate monohydrateManganous sulphate monohydrateManganese sulfate monohydrateManganese sulphate monohydrateManganese(II) sulphate hydrateManganese(II) sulfate hydrate

0248-500G
97061-302EA 65.6 USD InStock
97061-302
Manganese(II) sulfate monohydrate, Reagent Grade
Manganese (II) sulphate monohydrate


Formula: MnSO₄·1H₂O
MW: 169.02 g/mol
Melting Pt: 700 °C
Density: 2.95 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00149159
CAS Number: 10034-96-5
EINECS: 232-089-9
UN: 3077
ADR: 9,III
Merck Index: 13,05763
Order Now
Specification Test Results
Heavy Metals (as Pb) 0.05 %
Insolubles 0.08 %
Purity 98 %

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty