Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bovine Serum Albumin (BSA)

Supplier: Avantor
0332-100G VWRV0332-5KG 0332-500G VWRV0332-10KG 0332-25G 0332-1KG
97061-422CS 3929.59 USD InStock
97061-422 97061-420 80761-144 80761-134 97061-416 97061-418
Bovine Serum Albumin (BSA)
Proteins and Peptides
Bovine serum albumin, powder, Fraction V. Nuclease and protease-free. Suitable for use in most molecular biology applications.

  • Nuclease-free
  • Protease-free
  • Heavy Metals (as Pb) <0.001 %
  • Iron <0.0005 %
  • Purity >98 %

pH (5%, Water) @25 °C 6.5 - 7.5.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty