Synonyms:
Guanidinium sulfocyanide
, Guanidine Thiocyanate
, Guanidinium sulphocyanide
, GTC
, Guanidinium rhodanide
0380-500G
0380-250G
97061-522EA
257.38
USD
InStock
97061-522
97061-524
Guanidinium thiocyanate ≥99.0% for biotechnology
Guanidinium thiocyanate
A powerful protein denaturant used to inactivate endogenous RNases during the isolation of RNA from tissues and bacterial cultures