Synonyms:
Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulfonate)
, 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulphonic acid) diammonium salt
, Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulphonate)
, 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) diammonium salt
0400-10G
0400-1G
97061-576EA
415.86
USD
InStock
97061-576
97061-578
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulfonate])
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])
Substrate for the detection of horseradish peroxidase commonly used in ELISA. Read at 450 nm.
Soluble.Reaction Condition: 1 mM in 50 mM Sodium Citrate, 150 mM Sodium Phosphate and 0.01% H₂O₂. Stop with 1.5 mM NaN₃ in 0.1 M Citric Acid.